Anderson's Basal salt mixture
Anderson's basal salt mixture 10×1 L
Katalog # APOSPMA267A-10 X 1
New Product
Leverantör: Apollo Scientific
Anderson's Basal salt mixture
Katalog # APOSPMA267A-10 X 1
Leverantör: Apollo Scientific
New Product
CAS-nummer:
Gå till:
Product Details & Documents
Contains macro/micronutrients as described byAnderson, 1978/80 Plant tissue culture testedFull specification given in appendix.
- MFCD00243139
Specifikationer
Specifikationer
- Beskrivning:Anderson's basal salt mixture 10×1 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



