Orders LinkContactUsLinkComponent
Dispensing Lines
Dispensing Lines
Additional sets of dispensing lines, TUB-DOSIF-3,2, have an extra set to avoid stopping dispensing in case of contamination, cleaning, or breakage; 2 m length; 3,2 mm int.Ø, 6,4 mm ext.Ø
Katalog # ESPITUB-DOSIF-3.2
New Product
Leverantör:   RAYPA
Dispensing Lines
Katalog # ESPITUB-DOSIF-3.2
Leverantör:  RAYPA
New Product
Dispensing Lines
Dispensing Lines
Additional sets of dispensing lines, TUB-DOSIF-3,2, have an extra set to avoid stopping dispensing in case of contamination, cleaning, or breakage; 2 m length; 3,2 mm int.Ø, 6,4 mm ext.Ø
Price
Fråga efter pris
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
Additional silicone dosing tubes, with metal nozzle for dispensing sterile culture media.
  • Silicone dosing tubes with press-fit connection and metal nozzle
  • Sterile medium is dispensed through the flame-sterilizable nozzle
  • Made of MVQ silicone; heat-resistant up to 200 °C; 60 A hardness
  • Available in multiple diameters
  • Steam and pressurised air enable easy line cleaning before, during, and after use
  • All models are autoclavable, but autoclaving after each use is unnecessary thanks to the built-in steam cleaning program
Beställningsinformation: Dispensing lines can be customised, adjusting their length or adding modifications to adapt them to the specific production requirements of the customer.
Specifikationer
  • Beskrivning:
    Additional sets of dispensing lines, TUB-DOSIF-3,2, have an extra set to avoid stopping dispensing in case of contamination, cleaning, or breakage; 2 m length; 3,2 mm int.Ø, 6,4 mm ext.Ø
  • Seq:
    1