Temperature Data Loggers
Stainless steel and plastic connection base BDLDISK_CL, 120 g, for communication between the data logger and the software installed on a PC, 60×95×175 mm (External)
Catalog # ESPIBDLDISK_CL
New Product
Supplier: RAYPA
Jump to:
Product Details & Documents
AISI-316L stainless steel disk temperature recorder with connection base and software.
- User-friendly operation
- Data logger available in multiple sizes to suit different needs
- Delivers high accuracy and precision
Recommended for autoclave validation and for monitoring the internal temperature of containers.
Specifications
- Beskrivning:Stainless steel and plastic connection base BDLDISK_CL, 120 g, for communication between the data logger and the software installed on a PC
- Seq:1
- B×D×H:60×95×175 mm (External)