JONB CELLULOSA W CM52 2 KG 1 * 2 kg
Katalog # WHAT4037-200
Leverantör: Whatman products (Cytiva)
Frequently Bought Together
Förseglingsfilm för manuell applicering, Parafilm® M
Leverantör: HEATHROW
Aceton ≥99%, TECHNICAL
Leverantör: VWR Chemicals
VWR®, Serologisk pipett, standard
Leverantör: VWR Collection
Handskar, nitril, Kimtech™ Purple Nitrile™
Leverantör: KIMBERLY CLARK
VWR®, Ansiktsservett
Leverantör: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverantör: VWR Chemicals
VWR®, Nitrilhandskar
Leverantör: VWR Collection
Teststrips, MQuant®, Supelco®
Leverantör: Merck
VWR® NITRILE LIGHT, Nitrilhandskar
Leverantör: VWR Collection
VWR®, Transferpipetter, hög prestanda
Leverantör: VWR Collection
Metanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade för HPLC
Leverantör: VWR Chemicals
Vatten, HiPerSolv CHROMANORM® för HPLC
Leverantör: VWR Chemicals