O-Ring Replacement Kit
O-Ring replacement kit for bolts and threads, A27-8×50 Rotor
Katalog # SORV75007009
New Product
Leverantör: Thermo Fisher Scientific
Gå till:
Product Details & Documents
O-Ring replacement kit which includes grease for bolts and threads.
Specifikationer
- Beskrivning:O-Ring replacement kit for bolts and threads
- Kompatibel med:A27-8×50 Rotor
- Cat. No.:SORV75007009
- Material:Aluminium