Orders LinkContactUsLinkComponent
EdgePLUS Digital Microscopes
New Product
EdgePLUS Digital Microscopes
New Product
EdgePLUS Digital Microscopes
The microscope is designed to provide high fidelity colour reproduction with its low-light performance and flexible LED control features for observing even your target’s most fine details.
  • High quality optics
    1,3 megapixels
    Flexible LED control (FLC)
    Robust metallic housing
    Adjustable polariser
The optics adopted in the Edge series reveals the finest details, answering the needs of the most demanding microscopy applications. The advanced CMOS image sensor can transmit fluid and crisp images at 30 FPS with resolution up to 1280×960.
Tasking with software, the FLC maximises illumination flexibility by offering independent on/off control of the four LED quadrants in addition to the 6-levels intensity adjustment capability. The robust metal housing made of anodised alloy protects the Dino-lite from UV and electromagnetic interferences.
The built-in adjustable polariser allows to remove freely the unwanted reflection or glare from the object surface for a better contrast.
  • Catalog No:
  • IDCPAM4517MT-RFYW
  • IDCPAM4117MZT
  • IDCPAM4517MZT
  • IDCPAM4517MZTL
  • IDCPAM4917MT8
  • IDCPAM4917MZT4
  • Bildhastighet:
  • 30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEG
  • 30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEG
  • 30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEG
  • 30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEG
  • 30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEG
  • 30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEG
  • Datorkrav:
  • CPU: 2,5 GHz dual-core Intel Core i5 equivalent or greater, RAM: 4 GB or more, HD: 150 MB space available or more
  • CPU: 2,5 GHz dual-core Intel Core i5 equivalent or greater, RAM: 4 GB or more, HD: 150 MB space available or more
  • CPU: 2,5 GHz dual-core Intel Core i5 equivalent or greater, RAM: 4 GB or more, HD: 150 MB space available or more
  • CPU: 2,5 GHz dual-core Intel Core i5 equivalent or greater, RAM: 4 GB or more, HD: 150 MB space available or more
  • CPU: 2,5 GHz dual-core Intel Core i5 equivalent or greater, RAM: 4 GB or more, HD: 150 MB space available or more
  • Windows 7, 8, 10, 11, MacOS 10.12 and up
  • Programvara:
  • DinoCapture 2.0 (Windows), DinoXcope (Mac OS)
  • DinoCapture 2.0 (Windows), DinoXcope (Mac OS)
  • DinoCapture 2.0 (Windows), DinoXcope (Mac OS)
  • DinoCapture 2.0 (Windows), DinoXcope (Mac OS)
  • DinoCapture 2.0 (Windows), DinoXcope (Mac OS)
  • DinoCapture 2.0 (Windows), DinoXcope (Mac OS)
  • Vikt:
  • 90 g
  • 90 g
  • 90 g
  • 130 g
  • 130 g
  • 130 g
Product Family Options
  • Beskrivning
    Modell
    Förstoring
    Belysning
    Upplösning
  • Digital microscope, Ø: 32 mm, Height: 104 mm
    AM4517MT-RFYW
    20 - 220X
    White light, 8 LEDs
    1,3 Megapixel (1280×960)
    1 * 1 Items
    1 * 1 Items - 10 800,00 kr
    10 800,00 kr
    1 * 1 Items
       10 800,00 kr
    New Product
    Specifications
    Cat. No.
    IDCPAM4517MT-RFYW
    Bildhastighet
    30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEG
    Datorkrav
    CPU: 2,5 GHz dual-core Intel Core i5 equivalent or greater, RAM: 4 GB or more, HD: 150 MB space available or more
    Programvara
    DinoCapture 2.0 (Windows), DinoXcope (Mac OS)
    Vikt
    90 g
    New Product
  • Digital microscope, Ø: 33 mm, Height: 104 mm
    AM4117MZT
    10 - 220X
    White light, 8 LEDs
    1,3 Megapixel (1280×960)
    1 * 1 Items
    1 * 1 Items - 8 725,00 kr
    8 725,00 kr
    1 * 1 Items
       8 725,00 kr
    New Product
    Specifications
    Cat. No.
    IDCPAM4117MZT
    Bildhastighet
    30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEG
    Datorkrav
    CPU: 2,5 GHz dual-core Intel Core i5 equivalent or greater, RAM: 4 GB or more, HD: 150 MB space available or more
    Programvara
    DinoCapture 2.0 (Windows), DinoXcope (Mac OS)
    Vikt
    90 g
    New Product
  • Digital microscope, Ø: 33 mm, Length: 105 mm
    AM4517MZT
    10 - 220X
    White light, 8 LEDs
    1,3 Megapixel (1280×960)
    1 * 1 Items
    1 * 1 Items - 9 400,00 kr
    9 400,00 kr
    1 * 1 Items
       9 400,00 kr
    New Product
    Specifications
    Cat. No.
    IDCPAM4517MZT
    Bildhastighet
    30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEG
    Datorkrav
    CPU: 2,5 GHz dual-core Intel Core i5 equivalent or greater, RAM: 4 GB or more, HD: 150 MB space available or more
    Programvara
    DinoCapture 2.0 (Windows), DinoXcope (Mac OS)
    Vikt
    90 g
    New Product
  • Digital microscope, Ø: 33 mm, Length: 105 mm
    AM4517MZTL
    10 - 140X
    White light, 8 LEDs
    1,3 Megapixel (1280×960)
    1 * 1 Items
    1 * 1 Items - 9 400,00 kr
    9 400,00 kr
    1 * 1 Items
       9 400,00 kr
    New Product
    Specifications
    Cat. No.
    IDCPAM4517MZTL
    Bildhastighet
    30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEG
    Datorkrav
    CPU: 2,5 GHz dual-core Intel Core i5 equivalent or greater, RAM: 4 GB or more, HD: 150 MB space available or more
    Programvara
    DinoCapture 2.0 (Windows), DinoXcope (Mac OS)
    Vikt
    130 g
    New Product
  • Digital microscope, Ø: 33 mm, Height: 107 mm
    AM4917MT8
    700 - 900X
    White light, 8 LEDs
    1,3 Megapixel (1280×960)
    1 * 1 Items
    1 * 1 Items - 12 200,00 kr
    12 200,00 kr
    1 * 1 Items
       12 200,00 kr
    New Product
    Specifications
    Cat. No.
    IDCPAM4917MT8
    Bildhastighet
    30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEG
    Datorkrav
    CPU: 2,5 GHz dual-core Intel Core i5 equivalent or greater, RAM: 4 GB or more, HD: 150 MB space available or more
    Programvara
    DinoCapture 2.0 (Windows), DinoXcope (Mac OS)
    Vikt
    130 g
    New Product
  • Digital microscope, Ø: 33 mm, Height: 107 mm
    AM4917MZT4
    400 ~ 470x
    White light, 8 LEDs
    1,3 Megapixel (1280×960)
    1 * 1 Items
    1 * 1 Items - 12 200,00 kr
    12 200,00 kr
    1 * 1 Items
       12 200,00 kr
    New Product
    Specifications
    Cat. No.
    IDCPAM4917MZT4
    Bildhastighet
    30 fps at 1,3 MP, MJPEG; 30 fps at VGA, MJPEG
    Datorkrav
    Windows 7, 8, 10, 11, MacOS 10.12 and up
    Programvara
    DinoCapture 2.0 (Windows), DinoXcope (Mac OS)
    Vikt
    130 g
    New Product