Additional Peristaltic Pump
Additional peristaltic pump CAB-2, for applications like the production of test tubes in microbiology, where small to medium volumes are dispensed with each use
Katalog # ESPICAB-2
New Product
Leverantör: RAYPA
Gå till:
Product Details & Documents
This option is perfect for applications that require the dispensing of small to medium volumes, such as the production of test tubes in microbiology, the filling of Petri dishes, and other similar processes.
Included items: silicone tube, complementary head and Y-shaped tube
- To activate each dispensation, there are two available mechanisms: manual dispensing using a foot pedal and semi-automatic dispensing with delay
Depending on the combination of the chosen tube size and the number of installed peristaltic pumps, the dispensing speed in this mode ranges between 7 and 33 ml/s.
Specifikationer
- Beskrivning:Additional peristaltic pump CAB-2, for applications like the production of test tubes in microbiology, where small to medium volumes are dispensed with each use