SINGLE StEP® Empty Columns
SINGLE StEP® Empty fritted column; 10 each of 10, 25, 50, 100, and 200 ml, StEP, E Thomson InstrumenthotEP® EmptyMPNtEPsoG
Katalog # THMS94520-10
Leverantör: THOMSON INSTRUMENT
Gå till:
Product Details & Documents
Thomson Instrument Company is aware of the need to customise an available apparatus to individual experiments.
The SINGLE StEP® Empty columns allow users the opportunity to fill different sized columns with resins for purification applications. The wide range of columns offered means the chemist does not have be limited by column sizes. Empty columns also allow for the ability to connect these columns to FPLC systems.
Specifikationer
- Beskrivning:SINGLE StEP® Empty fritted column; 10 each of 10, 25, 50, 100, and 200 ml
- Seq:6
- Phase:StEP
- Packing material:E Thomson InstrumenthotEP® EmptyMPNtEPsoG