Filter Vial Toggle Press
Filter vial toggle press
Katalog # THMS35005
Leverantör: THOMSON INSTRUMENT
Gå till:
Product Details & Documents
The Thomson filter vial toggle press quickly depresses autosampler-ready filter vials.
Filter vial toggle press depresses five filter vials at one time
Specifikationer
- Beskrivning:Filter vial toggle press