EZ-Pak Filters
Katalog # MILXEZHABG504
Leverantör: Merck
New Product
Specifikationer
- Ø:50 mm
- Porstorlek:0,45 µm
- Beskrivning:EZ-Pak filters black, gridded
- Seq:2
Specifikationer
Om denna produkt
EZ-Pak membranes and dispenser revolutionise filter handling for MF method testing by combining the benefits of ready-to-use, sterile, individually-packed membranes with unique handling ease. From convenient, ready-to-use test kits to individual tools, simply select the items that are right for you.
Applications: Clinical, cosmetics, environmental, food and beverage, industrial QC, pharmaceutical