ContactUsLinkComponent
Tork S4 Hand Sanitising Alcohol-free Foam
Tork S4 Hand Sanitising Alcohol-free Foam
Catalog # SCAA520202
Supplier:  Essity
New Product
undefined
Tork S4 Hand Sanitising Alcohol-free Foam
Catalog # SCAA520202
Supplier:  Essity
New Product
Specifications
  • Beskrivning:
    Premium foam sanitiser, refill, 6× 1000 ml
Specifications
About this item
Tork hand sanitising alcohol-free foam S4, dermatologically tested formula: moisturising and gentle to the skin.
  • Help to protect guests - this hand sanitiser conforms with EN 1500 and EN 14476 and is effective against all enveloped viruses
  • Prove environmental commitment - this biodegradable hand sanitiser foam contains a 100% plant-based active ingredient
  • Be kind to skin - our alcohol-free hand sanitiser is dermatologically tested and contains moisturising ingredients
  • Meet safety standards - this non-flammable hand sanitiser has no CLP symbols, warning phrases or transport restrictions
Ensure good hygiene with our alcohol-free hand sanitising foam. This lactic acid formula is ideal for places where washroom facilities are unavailable and where alcohol-based products aren't an option.
It also contains a 100% plant-based active ingredient and is biodegradable, making it a more sustainable choice. Plus, it is effective against all enveloped viruses. Use with any S4 Tork dispenser.
Caution:
Use biocides safely. Always read the label and product information before use.