Tork Hand Sanitising Alcohol Foam
Katalog # SCAA590102
Leverantör: Essity
New Product
Specifikationer
- Beskrivning:Premium foam sanitiser, refill, 24× 47 ml
Specifikationer
Om denna produkt
Tork hand sanitising alcohol foam, gentle to the skin.
Conform with EN 1500 and EN 14476 (all enveloped, such as HIV, SARS Corona, HBV, HCV, Influenza H1N1 and H5N1) with this effective formula
- Provide a hand sanitiser that's gentle to the skin and easy to handle - this foam formula spreads easily
- Show that you care for user's skin, thanks to this alcohol hand sanitiser's dermatologically tested formula
Help prevent the spread of infections with Tork hand sanitising alcohol foam. Containing 75% w/w denatured alcohol, it is ideal for sanitising hands in offices and other spaces where waterless hand hygiene is required.
This effective formula is gentle and easy to apply to the skin and conforms with European Standard EN 1500 and EN 14476 (all enveloped viruses). Combine with our Tork Skincare dispenser, which is proven easy to use and provides good hand hygiene to all users.
Caution:
Use biocides safely. Always read the label and product information before use.