HPLC Columns, Reprospher 100 Biphenyl
Leverantör: DR MAISCH
New Product
Certifikat
Om denna produkt
Reprospher media are based on a 100 Å ultrapure silica and are fully scalable from UPLC (1,9 µm particles) to preparative and process scale applications.
Phase/modification: Biphenyl, Si-Phe-Phe
Specifikationer
- Catalog No:
- MAISRS15.BPE.S1520
- MAISRS15.BPE.S1503
- Endcapped:
- Dubbel
- Dubbel
- pH område:
- 2 - 8
- 2 - 8
- Surface area:
- 350 m²/g
- 350 m²/g