Orders LinkContactUsLinkComponent
ORCA Response Temperature-Controlled Packaging Solutions
New Product
ORCA Response Temperature-Controlled Packaging Solutions
New Product
ORCA Response Temperature-Controlled Packaging Solutions
ORCA Response is a multi-use, handheld, temperature-controlled packaging solution for transporting smaller payloads of red blood cells, platelets, pharmaceutical products and be easily carried and stored, making it the ideal solution for taking personal medicines on long journeys, including suitability as hand luggage on aeroplanes.
  • Easy to assemble
    Utilises Phase Change Material (PCM) and Vacuum Insulation Panel (VIP) technologies
    Universal pack out, Simple conditioning and assembly
    3 PCM temperature ranges: 2...8 ˚C, 15...25 ˚C, –20 ˚C
    Qualified for use with dry ice as coolant
    Optional carry bag for easy portability
    Optional ORCA Medic carry bag for military applications
    Suitable as hand luggage when flying
Advanced Phase Change Material (PCM) and Vacuum Insulation Panel (VIP) technologies ORCA response is qualified against ISTA 7D Summer and Winter temperature profiles, and enable 60+ hours of temperature protection.
Designed with the end user in mind ORCA Response combines high-performance technology and ease of use where rapid response packaging is required. This optimum performance and small volumetric weight make ORCA Response a clear choice for temperature-sensitive transportation and emergency applications.
Product Family Options
ORCA Response
  • Beskrivning
    B×D×H, utv
  • ORCA Response 2 L dry ice, 7,7 L payload volume, 1,9 kg
    347×260×223 mm
    1 * 1 Items
    1 * 1 Items - 1 800,00 kr
    1 800,00 kr
    1 * 1 Items
       1 800,00 kr
    New Product
    New Product
  • ORCA Response 2 L , –20 °C, 2,2 L payload volume, 6 kg
    347×260×223 mm
    1 * 1 Items
    1 * 1 Items - 2 925,00 kr
    2 925,00 kr
    1 * 1 Items
       2 925,00 kr
    New Product
    New Product
ORCA Response Carry Bag
  • Beskrivning
    B×D×H, utv
  • ORCA Response 2 L carry bag, with carry strap
    347×273×233 mm
    1 * 1 Items
    1 * 1 Items - 1 025,00 kr
    1 025,00 kr
    1 * 1 Items
       1 025,00 kr