ContactUsLinkComponent
PureClean™ Tyvek® 1421B Autoclavable Muff Covers
PureClean™ Tyvek® 1421B Autoclavable Muff Covers
Catalog # KACC10TK12
New Product
PureClean™ Tyvek® 1421B Autoclavable Muff Covers
Catalog # KACC10TK12
New Product
Specifications
  • Färg:
    Vit
  • Beskrivning:
    PureClean™ 1421B Tyvek® muff covers, fits 140 - 203 mm Ø opening
  • Seq:
    1
Specifications
About this item
PureClean™ Autoclavable muff covers are made from 1421B DuPont™ Tyvek®, which is a new grade of uncoated, untreated, flexible Tyvek® material. It has been specifically designed for pharmaceutical use in steam sterilisation applications. PureClean™ Tyvek® material does not have a water soluble antistat agent applied to it during its manufacturing process.
  • Uncoated Tyvek® for maximum breathability during sterilisation
  • Eliminate the risk of residue deposits on equipment post sterilisation
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labour costs and simplifies the conformance to wrapping SOP's
  • Autoclavable at 121 to 125 °C
  • Tear- and puncture-resistant, hydrophobic, lint-free
PureClean™ Tyvek® muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes, and hoses.
These covers are breathable and can be used to prevent particulates and microbial contamination from entering openings of any size. Since they are autoclavable, they can be used during sterilisation to reduce the time-consuming wrapping of open containers. Each closure incorporates latex-free rubber elastic to create a positive seal around round shaped openings.
Packaging:
Conveniently packaged, 100 per sealed poly bag.