Tork Clarity Foam Soap
Catalog # SCAA520201
Supplier: Essity
New Product
Specifications
- Förp.:6× 1000 ml
- Beskrivning:Premium foam soap, refill
Specifications
About this item
Tork clarity foam soap contains sustainble formula - 99% of ingredients are of natural origin.
35% lower water consumption* compared to Tork Mild Foam Soap (*Based on 2021 third-party verified LCA)
- Washing ingredients are from renewable sources (vegetable origins)
-
The formula is dermatologically tested, of a skin-friendly pH and readily biodegradable (based on third-party lab test according to OECD301B)
Perfume free fragrance, premium quality
Colour: Transparent
Compatible with S4 dispensers
Sustainability aspirations can be achieved without compromising hand hygiene with the new Tork clarity hand washing foam soap. It is formulated using a limited number of ingredients, 99% of which are from natural origins. This foam soap is suitable for Tork S4 dispensers, is certified easy-to-use and provides good hand hygiene to all users thanks to a factory-sealed refill bottle.
Certifications:
EU Ecolabel, SE/030/002.