Specifications
- Förp.:4 packs, 120 cloths each (4×120 cloths)
- Färg:Vit
- Beskrivning:Cloths, refill, premium, compatible with Tork W4 dispensers
- L×B:108×355 mm (Folded size), 415×355 mm (Un folded size)
Specifications
About this item
Multipurpose cleaning cloths for light-duty cleaning tasks. Use these durable cloths wet or dry to clean dust and dirt, and can rinse and reuse them as needed.
These disposable cleaning cloths are compatible with most chemical solvents, so no need to change the cleaning products
Compatible Top-pak dispenser has one-at-a-time dispensing to help optimise consumption and minimise waste
Tork light-duty cleaning cloths are an economical, multipurpose wiper. Compatible with most chemical solvents, these durable cloths can be used wet or dry for various light-duty cleaning tasks. They’re soft and flexible, making it easy to clean in hard-to-reach places, and more durable than a paper wiper, so can rinse and reuse them. The compatible Top-pak dispenser delivers one-at-a-time dispensing to minimise waste.