Millipore® Immobilon®-FL NOW Bundle Hydrophobic PVDF Transfer Membrane
Catalog # MILFNWKITFL
Supplier: Merck
New Product
Specifications
- Beskrivning:Immobilon®-FL NOW bundle transfer membrane
- Mått:85×10000 mm
Specifications
About this item
Hydrophobic PVDF transfer membrane for western blotting using fluorescent, chemifluorescence, chromogenic, or chemiluminescence.
Western blotting
Hydrophobic
Plain/white filter
0,45 µm pore size
Comes in a 8,5 cm × 10 m roll
The Immobilon NOW system offers 1-cut convenience for standard mini-blots (7 × 8,5cm) and midi-blots (8,5 × 13,5cm).
Compatible stains: All fluorescence dyes, Coomassie brilliant blue, Amido black, Ponceau-S red and Sypro® ruby