Millipore® Immobilon®-PSQ NOW Bundle Hydrophobic PVDF Transfer Membrane
Catalog # MILFNWKITSQ
Supplier: Merck
New Product
Specifications
- Beskrivning:Immobilon®-PSQ NOW bundle hydrophobic PVDF transfer membrane
- Mått:265×3750 mm
Specifications
About this item
PVDF transfer membrane hydrophobic for western blotting using chemiluminescence, chromogenic or radioactive detection methods.
Western blotting
Dot blot
White/plain filter
Electrostatic, hydrophobic
0,2 µm filter pore size
Comes in an 8,5 cm × 10 m roll
The Immobilon NOW system offers 1-cut convenience for standard mini-blots (7×8,5 cm) and midi-blots (8,5×13,5 cm).
Compatible stainn: Coomassie brilliant blue, Amido black, India ink, Ponceau-S red, Colloidal gold, CPTS, Toluidine blue, Transillumination and Sypro® ruby.