Undercounter Laboratory Refrigerators, TSU
Leverantör: Thermo Scientific
New Product
Certifikat
Om denna produkt
Thermo Scientific™ TSU series undercounter laboratory refrigerators offer a compelling cold storage option for a wide variety of laboratories. With a full suite of alarms, whisper quiet operation, and NSF 456 certified performance, the TSU series is designed specifically for laboratory settings.
Remote alarm contacts
- Whisper quiet operation
- Green refrigerants support global greenhouse gas emission reduction initiatives
- Hi/Lo temperature, door ajar, and power fail alarms
Certifications:
NSF 456 certified. ENERGYStar certified.
Specifikationer
- Catalog No:
- HERATSU405SV
- HERATSU405GV
- Temperatur:
- 2...8 °C
- 2...8 °C
- Avfrostning:
- Automatic
- Automatic
- Hyllor/lådor:
- 2 adjustable
- 2 adjustable
- Elektrisk:
- 230 V, 50 Hz
- 230 V, 50 Hz
- Nätsladd:
- EU
- EU
- Yttermått, B×D×H:
- 52×65×83 cm
- 52×65×83 cm