Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Nukleinsyrareagens
Nukleotider
TriLink® 2'-Deoxycytidine-5'-O-(1-Thiotriphosphate)
TriLink® 2'-Deoxycytidine-5'-O-(1-Thiotriphosphate)
:
TriLink BioTechnologies
New Product
TriLink® 2'-Deoxycytidine-5'-O-(1-Thiotriphosphate)
:
TriLink BioTechnologies
New Product
Grade: RUO
:
Item will be delivered at or below –20 °C and should be stored at or below –20 °C.