Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Nukleinsyrareagens
Nukleotider
TriLink® 5-Methyluridine-5'-Triphosphate
TriLink® 5-Methyluridine-5'-Triphosphate
TriLink® 5-Methyluridine-5'-Triphosphate
Leverantör:
TriLink BioTechnologies
New Product
Certifikat
Expandera bilden
TriLink® 5-Methyluridine-5'-Triphosphate
Leverantör:
TriLink BioTechnologies
New Product
Om denna produkt
Grade: RUO
Caution:
Item will be delivered at or below –20 °C and should be stored at or below –20 °C.