Order Entry
Sweden
ContactUsLinkComponent
Anti-ERK1+ERK2 (phospho T202) Rabbit Monoclonal Antibody [clone: S05-2H9]
Anti-ERK1+ERK2 (phospho T202) Rabbit Monoclonal Antibody [clone: S05-2H9]
Katalog # ABCAAB317377-100UL
Leverantör:  Abcam
New Product
undefined
Anti-ERK1+ERK2 (phospho T202) Rabbit Monoclonal Antibody [clone: S05-2H9]
Katalog # ABCAAB317377-100UL
Leverantör:  Abcam
New Product

Specifikationer

  • Storlek:
    100 µl
  • Förpackningstyp:
    Vial

Specifikationer

Om denna produkt

Rabbit monoclonal [S05-2H9] to ERK1 (phospho T202) + ERK2 (phospho T185).

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ERK1+ERK2
Clonality: Monoclonal
Clone: S05-2H9
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat; Chinese hamster