Anti-CD3 Rabbit Monoclonal Antibody [clone: SP162] (Alexa Fluor® 555)
ABCAAB307335-100
: Abcam
- Storlek:100 µl
- Förpackningstyp:Vial
Alexa Fluor® 555 Rabbit monoclonal [SP162] to CD3.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: SP162
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human