Anti-Metabotropic Glutamate Receptor 3/MGLUR3 Rabbit Monoclonal Antibody [clone: EPR9009(2)]
ABCAAB166608-100
: Abcam
- Storlek:100 µl
- Förpackningstyp:Vial
Rabbit monoclonal [EPR9009(2)] to Metabotropic Glutamate Receptor 3/MGLUR3.
Type: Primary
Antigen: Metabotropic Glutamate Receptor 3/MGLUR3
Clonality: Monoclonal
Clone: EPR9009(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Human; Rat