Order Entry
Sweden
ContactUsLinkComponent
Anti-Mad2L2/REV7 Rabbit Monoclonal Antibody [clone: EPR13657] (Alexa Fluor® 594)
Anti-Mad2L2/REV7 Rabbit Monoclonal Antibody [clone: EPR13657] (Alexa Fluor® 594)
Catalog # ABCAAB210455-100
Leverantör:  Abcam
undefined
Anti-Mad2L2/REV7 Rabbit Monoclonal Antibody [clone: EPR13657] (Alexa Fluor® 594)
Catalog # ABCAAB210455-100
Leverantör:  Abcam

Specifikationer

  • Seq:
    1286
  • Storlek:
    100 µl
  • Förpackningstyp:
    Vial

Specifikationer

Om denna produkt

Alexa Fluor® 594 Rabbit monoclonal [EPR13657] to Mad2L2/REV7.

Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Mad2L2/REV7
Clonality: Monoclonal
Clone: EPR13657
Conjugation: Alexa Fluor® 594
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human