Orders LinkContactUsLinkComponent
Adapters, receiver, with side socket, Quickfit®
Adapters, receiver, with side socket, Quickfit®
14/23 NS, 14/23 NS
Katalog # 271-0012
Leverantör:   SciLabware
Adapters, receiver, with side socket, Quickfit®
Katalog # 271-0012
Leverantör:  SciLabware
Adapters, receiver, with side socket, Quickfit®
Adapters, receiver, with side socket, Quickfit®
14/23 NS, 14/23 NS
Price
775,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Produktdetaljer och dokument
PYREX®, Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With side socket inclined at an angle of 105°.
Specifikationer
  • Kärna:
    14/23 NS
  • NS-hylsa:
    14/23 NS