ContactUsLinkComponent
VWR®, manipuleringssäker behållare för transport av biologiska prover
VWR®, manipuleringssäker behållare för transport av biologiska prover
Katalog # 720-1987
Leverantör:  VWR Collection
undefined
VWR®, manipuleringssäker behållare för transport av biologiska prover
Katalog # 720-1987
Leverantör:  VWR Collection
Specifikationer
  • Diam, övre:
    132 mm
  • Beskrivning:
    Seal tamper container
  • Höjd:
    161 mm
  • Förp.:
    120
Specifikationer
Om denna produkt
Container for transport of biological samples.
  • Disposable, with tamper-proof seal: Once closed the tube seal must be broken to access the contents
  • Leakproof to prevent accidental outflow of biological fluids that guarantees the safety of transportation personnel
  • Transparent and shock-proof for visual inspection of contents and for ensuring their integrity
Complete with test tube holder made of absorbent material holding 4×16 mm tubes, 4×12 mm tubes and 2×30 mm tubes at the same time, the test tubes are, therefore, protected if accidentally dropped, and fluids are absorbed in case of tube breakage.