Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Omrörare
Omrörningsverktyg
Magnetic stirring bar retriever, PTFE
Magnetic stirring bar retriever, PTFE
Magnetic stirring bar retriever, PTFE
Catalog # HECH42076010
Supplier:
GLASWARENFABRIK KARL HECHT
Expand image
Magnetic stirring bar retriever, PTFE
Catalog # HECH42076010
Supplier:
GLASWARENFABRIK KARL HECHT
Specifications
Ø:
8 mm
Längd:
350 mm
Förp.:
1
Specifications
About this item
Product Details
Product Details