Laboratory tray set
Tray, melamine, white, 350×250×40 mm
Katalog # HECH42000016
Leverantör: GLASWARENFABRIK KARL HECHT
Gå till:
Produktdetaljer och dokument
Complete laboratory tray set consists of plastic tray, Plexiglas® stand and 5 wide neck amber glass bottles (30 ml).
Specifikationer
- Beskrivning:Tray, melamine, white, 350×250×40 mm