Flasks acc. to Kjeldahl, with beaded rim
Katalog # HECH41870024
Leverantör: GLASWARENFABRIK KARL HECHT
Specifikationer
- Kapacitet:500 ml
- Förp.:1
Specifikationer
Om denna produkt
DURAN® borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With printed indication of contents and white labelling area.
Certifications: DIN 12360