Orders LinkContactUsLinkComponent
Reading magnifier
Reading magnifier
2,7×, 90×50 mm
Katalog # ESCH2668950
Leverantör:   ESCHENBACH OPTIK
Reading magnifier
Katalog # ESCH2668950
Leverantör:  ESCHENBACH OPTIK
Reading magnifier
Reading magnifier
2,7×, 90×50 mm
Price
1 400,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
This aplanatic magnifier is suitable for reading and looking at pictures for long periods.
  • Image free from spherical aberration
  • Edge-to-edge and distortion-free image
  • Comfortable thanks to the ergonomic handle
Specifikationer
  • Förstoring:
    2,7×
  • B×D:
    90×50 mm