Kylskåp, inbyggd, IK-serien
Leverantör: Liebherr
Certifikat
Om denna produkt
These IK series refrigerators are built-in appliances for integrated use.
- Ideal for integration into a fitted kitchen, creating a uniform look
- The unit door is fixed directly onto the appliance door
The unit door is hinged on the cabinet and connected to the appliance door with sliding rails. When the door is opened and closed, the unit door slides along the rails.