Solid phase extraction cartridges, Bond Elut™ C1
Leverantör: VARIAN
Certifikat
Om denna produkt
Due to the methyl group and subsequent low carbon load, Bond Elut™ C1 is the least retentive of all alkyl group bonded phases for non-polar compounds. However, due to the extensive end capping of this sorbent to mask polar silanol activity, retention and elution of polar and multi-functional analytes can still be achieved. Typical applications include plasma, urine and aqueous samples.
- Least retentive of all alkyl group bonded phases
- Easy retention and release of polar compounds
- Easy retention and release of multi-functional compounds
Specifikationer
- Catalog No:
- VARI12102004
- VARI12102090
- VARI12102031
- Kolbelastning:
- 4,1%
- 4,1%
- 4,1%
- Endcapped:
- Ja
- Ja
- Ja
- Surface area:
- 500 m²/g
- 500 m²/g
- 500 m²/g