L (+)-glutamin lösning 200 mM i 0,85% natriumklorid cell culture reagent
Katalog # LONZ17-605E
Supplier: LONZA
CAS Number:
Specifications
- Förpackningstyp:Plastic bottle
- Förp.:100 mL
- Seq:2440
Specifications
About this item
:
:146.15 g/mol
Merck Index:13,04484
Storage Temperature:Kylskåp
EINECS:200-292-1
CAS Number:56-85-9
Formula:C₅H₁₀N₂O₃
MDL Number:MFCD00008044