Perfluorodecahydronaphthalene (mixture of cis and trans isomers) ≥95.0%
TCIAP0837-25G
: TCI
:
- Förpackningstyp:Glass bottle
- Förp.:25 g
- Seq:3308
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 % (total of isomer)
:
:462.08
:14,04183
:142°C
:206-192-4
:306-94-5
:C₁₀F₁₈
:MFCD00010626