1-Nitropyrene ≥98.0%
Supplier: TCI
Certificates
About this item
CAS Number:5522-43-0
:
Formula:C₁₆H₉NO₂
MDL Number:MFCD00004138
:247.25 g/mol
Melting Pt:156 °C
EINECS:226-868-2
:GHS08
Storage Temperature:Kylskåp
Specification Test Results
- Appearance:Yellow to Amber powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:150.0 to 155.0 deg-C