Dietylkarbonat ≥98.0%
: TCI
:105-58-8
:
:C₅H₁₀O₃
:MFCD00009107
:118.13 g/mol
:14,03780
:126°C
:–43 °C
:0,9751 g/cm³ (20 °C)
:25°C
:Omgivande
:203-311-1
:2366
:3,III
:GHS02
:GHS07
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %