Safety spectacles, Horizon Flip Up
Katalog # DALL3024042-STD
Leverantör: HONEYWELL SAFETY
Specifikationer
- Typ:Abrasive resistant, shade lens colour IR 5
- Förp.:1
Specifikationer
Om denna produkt
The flip up dual lens safety glasses feature a clear impact resistant PC lens and a flip up shaded lens (IR 5).
- Ajustable temple wires and flip-up window
- Includes an integrated and superior sub-orbital protection and moulded sideshields
- Abrasive resistant
- Shade IR 5
Ideal for welders and industrial workers who require a shaded lens while working with high intensity light sources and a traditional clear impact-resistant polycarbonate lens when their task is complete.
Certifications: ANSI Z87.1-2003