Instrument trays with lid
Catalog # HECH42915003
Supplier: GLASWARENFABRIK KARL HECHT
Specifications
- B×D×H:200×100×50 mm
- Förp.:1
Specifications
About this item
Made of stainless steel.
- Resistant to hot-air sterilization up to +180 °C
- Excellent chemical resistance
- With knob lid