Test tube racks, wire
Catalog # HECH42815248
Supplier: GLASWARENFABRIK KARL HECHT
Specifications
- För rördiameter:12 mm
- Ant. hål:48 (4×12)
- Färg:Vit
- Höjd:40 mm
- Förp.:1
Specifications
About this item
Racks, made of plastic coated wire, ideally used for test tubes or centrifuge tubes.
- Good chemical resistance
- Good temperature resistance