Microtubes
without caps, conical, for autoanalyzer Olly, 1,5 ml
Katalog # KIMA18167
Leverantör: KIMA SAS DI CHIARIN RENZO
Gå till:
Product Details & Documents
PP, translucent.
- Without cap
- Excellent chemical resistance
- Excellent temperature resistance
Conical test tube for autoanalyser Olly type.
Specifikationer
- Kapacitet:1,5 ml
- Beskrivning:without caps, conical, for autoanalyzer Olly