Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(2-Pyridylazo)resorcinol
4-(2-Pyridylazo)resorcinol
4-(2-Pyridylazo)resorcinol
MOLEM40621555
:
Molekula
:
4-(2-Pyridylazo)resorcinol
MOLEM40621555
:
Molekula
:
Förpackningstyp:
Glass bottle for solids
Förp.:
5 g
Seq:
3173
:
:
215.21 g/mol
:
214-528-6
:
1141-59-9
:
C₁₁H₉N₃O₂
:
MFCD00044237