Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Flourescein dinatriumsalt
Flourescein dinatriumsalt
Flourescein dinatriumsalt
Flourescein dinatriumsalt
EHERC13750000
:
EHRENSTORFER
:
Flourescein dinatriumsalt
EHERC13750000
:
EHRENSTORFER
:
Förpackningstyp:
Glass bottle for solids
Förp.:
0,1 g
Seq:
3020
:
:
376.28 g/mol
:
13,04185
:
> 300 °C
:
1,601 g/cm³ (20 °C)
:
Omgivande
:
208-253-0
:
518-47-8
:
C₂₀H₁₀Na₂O₅
:
MFCD00167039