Bottles, wide neck, amber, with screw caps
WITG5.895.002
- Gänga:GL 80
- Kapacitet:2500 ml
- Seq:02
- Förp.:1
Amber bottles, wide mouth with DIN.
- Good chemical and physical properties
- For applications that do not involve high thermal stresses
- Round thread in accordance with DIN 168
Ideal for sampling, storing and transporting powders and pastes. Amber coloured to protect light-sensitive materials.