Desiccators, NOVUS
Catalog # WITG5.660.200
Supplier: witeg Labortechnik
Specifications
- Ø inr.:200 mm
- Typ:NOVUS, with needle valve stopcock, cone ST 24/29
- Seq:01
- Förp.:1
Specifications
About this item
This desiccator is made from DURAN® glass, without porcelain disc, with tubulated lid.