Harnesses, Ropax
Catalog # 113-1274
Supplier: HONEYWELL SAFETY
Specifications
- Fästpunkter:4-point
- Beskrivning:Harness, Polyester/stainless steel
- Storlek:L/XL
- Förp.:1
Specifications
About this item
This harness is ideal for industrial rope access, rescue and special forces. It can be used in conjunction with a climber's seat and an ascender.
- Black polyester webbing and stainless steel components offer optimum durability, chemical and abrasion resistance
- Triangular Suspension position splits the forces between the leg straps and the waist belt across four points of pressure
- Detachable webbing sling on the chest straps positions the ascender correctly in a vertical position to facilitate ascension
EN 358, EN 813, EN 361
Certifications: EN 358, EN 813, EN 361