VWR®, Bottle-Top Vacuum Filtration Systems, PES
Supplier: VWR Collection
Certificates
About this item
PS funnel and collection reservoir. Systems for vacuum filtration of aqueous solutions including cell culture media, buffers or other biological fluids.
- Each individual unit is lot numbered for traceability
- Once filtration is complete, solution can be stored in the collection reservoir until needed
- Non pyrogenic, non cytotoxic
PES membrane: Low protein binding and low extractables. Ideal for tissue culture applications.
0,2 µm: Ideal for sterilisation applications and media preparation.
0,45 µm: Ideal for buffer clarification.
Non pyrogenic, non cytotoxic
Certifications: ISO 10993-5: 1999.
Delivery: Full unit includes the filtration funnel with either 0,2 or 0,45 µm PES membrane, vacuum port, lid, collection reservoir and cap for storage.
Packaging: Individually packed in easy peel-to-open bags, receiver bottle cap is individually wrapped.
Specifications
- Catalog No:
- 514-0334
- 514-0345
- 514-0344
- 514-0329
- 514-0340
- 514-0328
- 514-0339
- 514-0331
- 514-0342
- 514-0330
- 514-0341
- 514-0336
- 514-0347
- 514-0335
- 514-0346
- 514-0338
- 514-0337
- 514-0333
- 514-0332
- 514-0343
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Miljöcertificering:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
Specifications
Related Information
Certificates
Search by Lot NumberAvantor Cell Culture Solutions.pdf VWR® for filtration
Product Literature
CoC VWR® Bottle-Top Vacuum Filtration Systems, PES.pdf
Conformity declaration
CoC_VWRI514-0328_190408053 CoC_VWRI514-0328_191022053 CoC_VWRI514-0328_200710053 CoC_VWRI514-0328_201010053 CoC_VWRI514-0328_210127053
More