Alcohol swab
Katalog # HERE1131897
Leverantör: HERENZ HEINZ
Specifikationer
- Beskrivning:Svabb, isopropylalkoholl, 30×30 mm
- Förp.:2.000
Specifikationer
Om denna produkt
70% isopropyl alcohol non-sterile swabs for cleaning applications.
Unfolded size of the swab is approximately 60×30 mm.
Packaging: Individual – 100 pieces per box.