Order Entry
Sweden
ContactUsLinkComponent
Iminodiacetic acid för syntes, Sigma-Aldrich®
Irritant
Iminodiacetic acid för syntes, Sigma-Aldrich®
Catalog # 8.14165.0250
Supplier:  Merck
CAS Number:  
Irritant
Iminodiacetic acid för syntes, Sigma-Aldrich®
Catalog # 8.14165.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Förpackningstyp:
    Plastic bottle
  • Förp.:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Solubility:
    42 g/l (20 °C)
  • Melting Point:
    247 °C (decomposition)
  • Molar Mass:
    133.10 g/mol
  • pH:
    2.2 - 2.3 (H2O, 20 °C) (saturated solution)
  • Hill Formula:
    C4H7NO4
  • Chemical Formula:
    HN(CH2COOH)2

Specification Test Results

About this item

 :
 :133.10 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:205-555-4
 :GHS07
Storage Temperature:Omgivande
CAS Number:142-73-4
Formula:C₄H₇NO₄
MDL Number:MFCD00004280